Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Brix1
Entrez Gene ID
294799
Gene Name
BRX1, biogenesis of ribosomes
Gene Aliases
Bxdc2, RGD1308508
Chromosome Location
Chr.2: 60450547 - 60461425 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029915.1 NP_001025086.1 5 - 6 427 88
GenBank mRNA BC098005.1 - 5 - 6 427 88

View Full Product Details