The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212532.2 | NP_997697.1 | 1 - 2 | 189 | 86 |
XM_008772715.1 | XP_008770937.1 | 1 - 2 | 451 | 86 | |
GenBank mRNA | BC097930.1 | - | 1 - 2 | 179 | 86 |
CO385374.1 | - | 1 - 2 | 189 | 86 |