Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ly6g6e
Entrez Gene ID
406866
Gene Name
lymphocyte antigen 6 complex, locus G6E
Gene Aliases
-
Chromosome Location
Chr.20: 5064279 - 5069032 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001972.1 NP_001001972.1 1 - 2 117 88
XM_006256071.2 XP_006256133.1 3 - 4 293 88
XM_006256072.2 XP_006256134.1 4 - 5 405 88

View Full Product Details