Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stk19
Entrez Gene ID
361800
Gene Name
serine/threonine kinase 19
Gene Aliases
-
Chromosome Location
Chr.20: 4317440 - 4517128 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013197.2 NP_001013215.2 4 - 5 518 67
GenBank mRNA BC061571.1 - 4 - 5 522 67

Gene Symbol
LOC103694374
Entrez Gene ID
103694374
Gene Name
serine/threonine-protein kinase 19
Gene Aliases
-
Chromosome Location
Chr.20: 2665615 - 2666517 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772703.1 XP_008770925.1 - 522 67

View Full Product Details