The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012468.1 | NP_001012486.1 | 4 - 5 | 306 | 151 |
XM_006254132.2 | XP_006254194.1 | 4 - 5 | 363 | 151 | |
GenBank mRNA | BC089957.1 | - | 4 - 5 | 306 | 151 |