Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
T
Entrez Gene ID
292301
Gene Name
brachyury
Gene Aliases
-
Chromosome Location
Chr.1: 52887067 - 52897323 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106209.1 NP_001099679.1 3 - 4 719 84
XM_006227918.2 XP_006227980.1 5 - 6 1619 84
XM_008758750.1 XP_008756972.1 2 - 3 868 84

View Full Product Details