Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc5a7
Entrez Gene ID
85426
Gene Name
solute carrier family 5 (sodium/choline cotransporter), member 7
Gene Aliases
Cht1
Chromosome Location
Chr.9: 5293381 - 5330976 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053521.1 NP_445973.1 2 - 3 399 72
XM_006244251.2 XP_006244313.1 2 - 3 502 72
XM_008766774.1 XP_008764996.1 2 - 3 550 72
XM_008766775.1 XP_008764997.1 2 - 3 499 72
GenBank mRNA AB030947.1 - 2 - 3 399 72

View Full Product Details