Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cstf1
Entrez Gene ID
311670
Gene Name
cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa
Gene Aliases
-
Chromosome Location
Chr.3: 170380414 - 170392162 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013161.1 NP_001013179.1 2 - 3 233 110
XM_006235681.1 XP_006235743.1 2 - 3 338 110
XM_006235682.1 XP_006235744.1 2 - 3 282 110
XM_006235683.2 XP_006235745.1 2 - 3 365 110
XM_006235684.2 XP_006235746.1 2 - 3 362 110
XM_006235685.2 XP_006235747.1 2 - 3 340 110
XM_006235686.2 XP_006235748.1 2 - 3 337 110
XM_006235687.2 XP_006235749.1 2 - 3 334 110
GenBank mRNA BC091381.1 - 2 - 3 233 110

View Full Product Details