Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rfx7
Entrez Gene ID
315804
Gene Name
regulatory factor X, 7
Gene Aliases
Rfxdc2
Chromosome Location
Chr.8: 79159292 - 79279962 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127490.1 NP_001120962.1 4 - 5 984 123
XM_006243326.2 XP_006243388.1 5 - 6 897 123
XM_008766291.1 XP_008764513.1 6 - 7 861 123
XM_008766292.1 XP_008764514.1 6 - 7 545 123
GenBank mRNA BC158676.1 - 4 - 5 415 123

View Full Product Details