Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Npc1
Entrez Gene ID
266732
Gene Name
Niemann-Pick disease, type C1
Gene Aliases
Cdig2
Chromosome Location
Chr.18: 3616878 - 3662715 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153624.2 NP_705888.2 12 - 13 2041 96
XM_006254403.2 XP_006254465.1 12 - 13 2097 96

View Full Product Details