Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrps18b
Entrez Gene ID
294230
Gene Name
mitochondrial ribosomal protein S18B
Gene Aliases
Ptd017
Chromosome Location
Chr.20: 3344870 - 3350929 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212534.2 NP_997699.2 6 - 7 498 87
GenBank mRNA BC166875.1 - 6 - 7 480 87
FQ215273.1 - 6 - 7 471 87

View Full Product Details