Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mdc1
Entrez Gene ID
309595
Gene Name
mediator of DNA damage checkpoint 1
Gene Aliases
-
Chromosome Location
Chr.20: 3405017 - 3419831 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166275.1 NP_001159747.1 2 - 3 307 83
GenBank mRNA BC085955.1 - 2 - 3 307 83

View Full Product Details