Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Anxa9
Entrez Gene ID
689830
Gene Name
annexin A9
Gene Aliases
-
Chromosome Location
Chr.2: 196470318 - 196481580 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001072193.5 XP_001072193.3 7 - 8 1743 112
XM_006232937.2 XP_006232999.1 7 - 8 1743 112

View Full Product Details