Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr18
Entrez Gene ID
314617
Gene Name
WD repeat domain 18
Gene Aliases
-
Chromosome Location
Chr.7: 12561318 - 12569120 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039027.1 NP_001034116.1 4 - 5 609 91
XM_006240906.1 XP_006240968.1 4 - 5 686 91
GenBank mRNA BC099828.1 - 4 - 5 609 91
FQ212635.1 - 4 - 5 612 91
FQ212816.1 - 4 - 5 612 91
FQ219525.1 - 4 - 5 612 91

View Full Product Details