Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr18
Entrez Gene ID
314617
Gene Name
WD repeat domain 18
Gene Aliases
-
Chromosome Location
Chr.7: 12561318 - 12569120 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039027.1 NP_001034116.1 9 - 10 1177 57
XM_006240906.1 XP_006240968.1 9 - 10 1254 57
GenBank mRNA BC099828.1 - 9 - 10 1177 57
FQ212816.1 - 9 - 10 1180 57
FQ219525.1 - 9 - 10 1181 57

View Full Product Details