Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nsfl1c
Entrez Gene ID
83809
Gene Name
NSFL1 (p97) cofactor (p47)
Gene Aliases
XY40, p47
Chromosome Location
Chr.3: 146980923 - 147005475 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031981.2 NP_114187.1 1 - 2 216 73
XM_006235254.2 XP_006235316.1 1 - 2 183 73
GenBank mRNA AB002086.1 - 1 - 2 216 73
BC072464.1 - 1 - 2 113 73
FQ228953.1 - 1 - 2 148 73
Y10769.1 - 1 - 2 113 73

View Full Product Details