Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dok4
Entrez Gene ID
361364
Gene Name
docking protein 4
Gene Aliases
-
Chromosome Location
Chr.19: 10563390 - 10575109 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108438.2 NP_001101908.1 1 - 2 150 92
XM_008772315.1 XP_008770537.1 1 - 2 183 92
GenBank mRNA BC158749.1 - 1 - 2 150 92

View Full Product Details