Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dok4
Entrez Gene ID
361364
Gene Name
docking protein 4
Gene Aliases
-
Chromosome Location
Chr.19: 10563390 - 10575109 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108438.2 NP_001101908.1 8 - 9 1156 101
XM_008772314.1 XP_008770536.1 6 - 7 1248 101
XM_008772315.1 XP_008770537.1 7 - 8 1340 101
XM_008772316.1 XP_008770538.1 7 - 8 1337 101
XM_008772317.1 XP_008770539.1 7 - 8 1096 101
GenBank mRNA BC158749.1 - 8 - 9 1156 101

View Full Product Details