The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107253.1 | NP_001100723.1 | 1 - 2 | 73 | 73 |
XM_006251763.2 | XP_006251825.1 | 2 - 3 | 1708 | 73 |