Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csn1s1
Entrez Gene ID
24284
Gene Name
casein alpha s1
Gene Aliases
Casa, Csn1, Csna
Chromosome Location
Chr.14: 21993450 - 22009308 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138874.2 NP_620229.2 18 - 19 573 61
GenBank mRNA J00710.1 - 18 - 19 573 61

View Full Product Details