Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4a8
Entrez Gene ID
266674
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 8
Gene Aliases
Cyp4a12
Chromosome Location
Chr.5: 133978953 - 134008255 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031605.2 NP_113793.2 8 - 9 1095 80
GenBank mRNA BC081771.1 - 8 - 9 1095 80
M37828.1 - 8 - 9 1085 80

View Full Product Details