Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc7a8
Entrez Gene ID
84551
Gene Name
solute carrier family 7 (amino acid transporter light chain, L system), member 8
Gene Aliases
Lat2, Lat4
Chromosome Location
Chr.15: 33369245 - 33428942 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053442.1 NP_445894.1 2 - 3 961 124
GenBank mRNA AB024400.1 - 2 - 3 961 124

View Full Product Details