Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl15
Entrez Gene ID
295985
Gene Name
methyltransferase like 15
Gene Aliases
RGD1304978
Chromosome Location
Chr.3: 100186106 - 100366148 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234687.2 XP_006234749.1 1 - 2 103 144

View Full Product Details