Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gca
Entrez Gene ID
295647
Gene Name
grancalcin
Gene Aliases
-
Chromosome Location
Chr.3: 48625991 - 48662778 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106483.2 NP_001099953.1 6 - 7 664 61

View Full Product Details