Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pot1
Entrez Gene ID
500054
Gene Name
protection of telomeres 1
Gene Aliases
Pot1a
Chromosome Location
Chr.4: 51888632 - 51946764 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024322.1 NP_001019493.1 9 - 10 1134 75
XM_006236160.2 XP_006236222.1 10 - 11 1226 75
XM_006236161.2 XP_006236223.1 11 - 12 1448 75
XM_008762809.1 XP_008761031.1 11 - 12 1630 75
XM_008762810.1 XP_008761032.1 11 - 12 1575 75
XM_008762811.1 XP_008761033.1 9 - 10 1179 75
GenBank mRNA BC085834.1 - 9 - 10 1134 75
BC090024.1 - 4 - 5 550 75

View Full Product Details