Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clec12a
Entrez Gene ID
680338
Gene Name
C-type lectin domain family 12, member A
Gene Aliases
-
Chromosome Location
Chr.4: 163112100 - 163130979 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134716.1 NP_001128188.1 - 385 148
GenBank mRNA BC168186.1 - - 385 148
GU357484.1 - - 184 148

View Full Product Details