Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pramef20
Entrez Gene ID
691150
Gene Name
PRAME family member 20
Gene Aliases
pramef25
Chromosome Location
Chr.5: 162406599 - 162411066 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001077012.1 XP_001077012.1 2 - 3 865 83

View Full Product Details