Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akirin1
Entrez Gene ID
595134
Gene Name
akirin 1
Gene Aliases
-
Chromosome Location
Chr.5: 141415226 - 141430627 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030054.1 NP_001025225.1 1 - 2 317 108
GenBank mRNA CV107792.1 - 1 - 2 317 108

View Full Product Details