The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177685.1 | NP_001171156.1 | 7 - 8 | 674 | 60 |
GenBank mRNA | BC162007.1 | - | 7 - 8 | 674 | 60 |