Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wipi1
Entrez Gene ID
303630
Gene Name
WD repeat domain, phosphoinositide interacting 1
Gene Aliases
RGD1307754
Chromosome Location
Chr.10: 97859730 - 97896886 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127297.1 NP_001120769.1 6 - 7 667 65
XM_008768366.1 XP_008766588.1 6 - 7 1286 65
XM_008768367.1 XP_008766589.1 6 - 7 564 65
GenBank mRNA BC166829.1 - 6 - 7 667 65

View Full Product Details