Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ergic1
Entrez Gene ID
287177
Gene Name
endoplasmic reticulum-golgi intermediate compartment (ERGIC) 1
Gene Aliases
-
Chromosome Location
Chr.10: 16821393 - 16897165 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767628.1 XP_008765850.1 8 - 9 595 94

View Full Product Details