Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfx
Entrez Gene ID
367832
Gene Name
zinc finger protein X-linked
Gene Aliases
RGD1562664
Chromosome Location
Chr.X: 63158368 - 63233899 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109017.1 NP_001102487.1 5 - 6 1241 87
XM_006257030.2 XP_006257092.1 5 - 6 964 87
XM_006257031.2 XP_006257093.1 4 - 5 1244 87
XM_006257032.2 XP_006257094.1 5 - 6 1311 87
XM_008773264.1 XP_008771486.1 4 - 5 896 87
XM_008773265.1 XP_008771487.1 5 - 6 1345 87

View Full Product Details