Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lamp3
Entrez Gene ID
303801
Gene Name
lysosomal-associated membrane protein 3
Gene Aliases
-
Chromosome Location
Chr.11: 84930940 - 85000694 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012015.1 NP_001012015.1 4 - 5 1022 96
XM_006248595.2 XP_006248657.1 8 - 9 1411 96
XM_008768886.1 XP_008767108.1 9 - 10 1711 96
XM_008768887.1 XP_008767109.1 5 - 6 1019 96
GenBank mRNA BC083787.1 - 4 - 5 1022 96

View Full Product Details