Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpd52l2
Entrez Gene ID
296480
Gene Name
tumor protein D52-like 2
Gene Aliases
-
Chromosome Location
Chr.3: 176989653 - 177009425 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235754.2 XP_006235816.1 6 - 7 655 68
XM_006235757.2 XP_006235819.1 5 - 6 615 68
XM_008762499.1 XP_008760721.1 6 - 7 589 68

View Full Product Details