Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xylt1
Entrez Gene ID
64133
Gene Name
xylosyltransferase 1
Gene Aliases
XYLT-1, Xt1
Chromosome Location
Chr.1: 186939698 - 187264758 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006230151.2 XP_006230213.2 12 - 13 2648 65
GenBank mRNA AJ295748.1 - 9 - 10 2154 65

View Full Product Details