Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xylt1
Entrez Gene ID
64133
Gene Name
xylosyltransferase 1
Gene Aliases
XYLT-1, Xt1
Chromosome Location
Chr.1: 186939698 - 187264758 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006230151.2 XP_006230213.2 3 - 4 491 59

View Full Product Details