Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dazl
Entrez Gene ID
680486
Gene Name
deleted in azoospermia-like
Gene Aliases
-
Chromosome Location
Chr.9: 12633990 - 12651039 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109414.1 NP_001102884.1 8 - 9 884 131
XM_006244477.2 XP_006244539.1 9 - 10 788 131

View Full Product Details