Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rftn1
Entrez Gene ID
501095
Gene Name
raftlin lipid raft linker 1
Gene Aliases
2310015n21rik, RGD1310777, RGD1563347
Chromosome Location
Chr.9: 12740885 - 12942358 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135011.1 NP_001128483.1 7 - 8 1305 92
XM_008766876.1 XP_008765098.1 7 - 8 1869 92
GenBank mRNA BC098817.1 - 1 - 2 40 92

View Full Product Details