Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Prl3d1
- Entrez Gene ID
-
53950
- Gene Name
- Prolactin family 3, subfamily d, member 1
- Gene Aliases
-
Csh1, Pl-I, Pl-Im, Pl1, Rhco1, rPl-1
- Chromosome Location
-
Chr.17: 39777255 - 39782877 on Build Rnor_6.0
- Gene Symbol
- LOC103690150
- Entrez Gene ID
-
103690150
- Gene Name
- prolactin-3D1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 39721993 - 39728391 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FQ228618.1 |
- |
- |
540 |
80 |
- Gene Symbol
- Prl3d2
- Entrez Gene ID
-
691552
- Gene Name
- Prolactin family 3, subfamily d, member 2
- Gene Aliases
-
Pl-1
- Chromosome Location
-
Chr.17: 42849469 - 42855494 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
D21103.1 |
- |
4 - 5 |
479 |
80 |
View Full Product Details