Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trub2
Entrez Gene ID
366012
Gene Name
TruB pseudouridine (psi) synthase family member 2
Gene Aliases
-
Chromosome Location
Chr.3: 8338479 - 8348746 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014257.2 NP_001014279.1 2 - 3 287 82
GenBank mRNA BC084712.1 - 2 - 3 262 82
CB613146.1 - 2 - 3 287 82
FQ225796.1 - 2 - 3 131 82
FQ231048.1 - 2 - 3 258 82
FQ231592.1 - 2 - 3 301 82

View Full Product Details