Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trub2
Entrez Gene ID
366012
Gene Name
TruB pseudouridine (psi) synthase family member 2
Gene Aliases
-
Chromosome Location
Chr.3: 8338479 - 8348746 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014257.2 NP_001014279.1 3 - 4 354 79
GenBank mRNA BC084712.1 - 3 - 4 329 79
CB613146.1 - 3 - 4 354 79
FQ225796.1 - 3 - 4 198 79
FQ231048.1 - 3 - 4 325 79
FQ231592.1 - 3 - 4 368 79

View Full Product Details