Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdk20
Entrez Gene ID
364666
Gene Name
cyclin-dependent kinase 20
Gene Aliases
Ccrk
Chromosome Location
Chr.17: 1936163 - 1942867 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025752.2 NP_001020923.2 2 - 3 403 63
GenBank mRNA BC098838.1 - 2 - 3 403 63

View Full Product Details