Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk20
Entrez Gene ID
364666
Gene Name
cyclin-dependent kinase 20
Gene Aliases
Ccrk
Chromosome Location
Chr.17: 1936163 - 1942867 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025752.2 NP_001020923.2 6 - 7 894 75
GenBank mRNA BC098838.1 - 5 - 6 1459 75

View Full Product Details