Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ntn4
Entrez Gene ID
299737
Gene Name
netrin 4
Gene Aliases
RGD1565947
Chromosome Location
Chr.7: 34533543 - 34649330 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106780.1 NP_001100250.1 6 - 7 1561 132

View Full Product Details