The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138526.1 | NP_612535.1 | 2 - 3 | 678 | 77 |
GenBank mRNA | AF199495.1 | - | 1 - 2 | 235 | 77 |