Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syce1l
Entrez Gene ID
100361431
Gene Name
synaptonemal complex central element protein 1-like
Gene Aliases
-
Chromosome Location
Chr.19: 45946430 - 45957115 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006255676.2 XP_006255738.1 7 - 8 445 82

View Full Product Details