Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbx3
Entrez Gene ID
297093
Gene Name
chromobox homolog 3
Gene Aliases
Hp1-gamma
Chromosome Location
Chr.4: 81243795 - 81257898 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008313.2 NP_001008314.2 4 - 5 406 78
XM_006236484.2 XP_006236546.1 4 - 5 1114 78
XM_006236485.2 XP_006236547.1 4 - 5 734 78
GenBank mRNA BC086601.1 - 4 - 5 406 78
BC127508.1 - 4 - 5 421 78

View Full Product Details