Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Filip1l
Entrez Gene ID
304020
Gene Name
filamin A interacting protein 1-like
Gene Aliases
RGD1565927
Chromosome Location
Chr.11: 45039488 - 45127820 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248213.2 XP_006248275.2 1 - 2 4476 100

View Full Product Details