The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007735.1 | NP_001007736.1 | 1 - 2 | 115 | 64 |
GenBank mRNA | BC061808.1 | - | 1 - 2 | 115 | 64 |
FQ214111.1 | - | 1 - 2 | 129 | 64 |