The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031643.1 | NP_001026813.1 | 10 - 11 | 1112 | 128 |
GenBank mRNA | BC098700.1 | - | 10 - 11 | 1112 | 128 |